siRNA Details
| Gene Symbol: CTSV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201575.1 | NP_001188504.1 | 6 | 691 | ||
| NM_001333.3 | NP_001324.2 | 6 | 894 | |||
| GenBank mRNA | AB001928.1 | - | 6 | 717 | ||
| AF070448.1 | - | 5 | 671 | |||
| AK310740.1 | - | 6 | 826 | |||
| AK312806.1 | - | 6 | 691 | |||
| AY358641.1 | - | 6 | 894 | |||
| BC023504.2 | - | 6 | 708 | 3139314 | ||
| BC110512.1 | - | 6 | 763 | 40031744 | ||
| Y14734.1 | - | 6 | 713 | |||

CTSV
Genomic Map