siRNA Details
| Gene Symbol: CTSO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001334.2 | NP_001325.1 | 5 | 705 | ||
| GenBank mRNA | AK300112.1 | - | 3 | 328 | ||
| AK308323.1 | - | 5 | 711 | |||
| AK312354.1 | - | 5 | 670 | |||
| BC049206.1 | - | 5 | 658 | 5189657 | ||
| X77383.1 | - | 5 | 705 | |||

CTSO
Genomic Map