siRNA Details
| Gene Symbol: FAM49A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030797.3 | NP_110424.1 | 10, 11 | 1085 | ||
| XM_005262630.3 | XP_005262687.1 | 9, 10 | 1041 | |||
| XM_006712108.3 | XP_006712171.1 | 10, 11 | 1040 | |||
| GenBank mRNA | AK001942.1 | - | 2, 3 | 141 | ||
| AK055334.1 | - | 10, 11 | 1053 | |||
| AK091280.1 | - | 5, 6 | 499 | |||
| AK315065.1 | - | 9, 10 | 980 | |||
| AL136704.1 | - | 10, 11 | 1050 | |||
| BC038971.1 | - | 10, 11 | 1036 | 5200807 | ||
| CR533504.1 | - | 8, 9 | 832 | |||

FAM49A
Genomic Map