siRNA Details
| Gene Symbol: ANKRD13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030816.4 | NP_110443.3 | 1 | 437 | ||
| XM_005271234.3 | XP_005271291.1 | 1 | 680 | |||
| XM_005271235.3 | XP_005271292.1 | 1 | 427 | |||
| XM_006710929.3 | XP_006710992.1 | 1 | 450 | |||
| GenBank mRNA | AL136717.1 | - | 1 | 438 | ||
| BC016937.2 | - | 1 | 419 | |||
| BC028840.1 | - | 1 | 426 | 4558748 | ||

ANKRD13C
Genomic Map