siRNA Details
| Gene Symbol: GFOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243650.1 | NP_001230579.1 | 2 | 479 | ||
| NM_030819.3 | NP_110446.3 | 2 | 479 | |||
| XM_017023738.1 | XP_016879227.1 | 2 | 429 | |||
| GenBank mRNA | AK074382.1 | - | 3 | 432 | ||
| AK225997.1 | - | 2 | 449 | |||
| AK307280.1 | - | 2 | 433 | |||
| AY358169.1 | - | 1 | 189 | |||
| BC002943.1 | - | 1 | 106 | |||
| BC107585.1 | - | 2 | 429 | 6463662 | ||
| DC368734.1 | - | 2 | 479 | |||
| DC381447.1 | - | 2 | 434 | |||

GFOD2
Genomic Map