siRNA Details
| Gene Symbol: URM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135947.2 | NP_001129419.1 | 2 | 127 | ||
| NM_001265582.1 | NP_001252511.1 | 2 | 127 | |||
| NM_030914.3 | NP_112176.1 | 2 | 127 | |||
| NR_049743.1 | 2 | 168 | ||||
| GenBank mRNA | AK001880.1 | - | 2 | 76 | ||
| AK097029.1 | - | 2 | 73 | |||
| AK300877.1 | - | 2 | 74 | |||
| BC003581.1 | - | 2 | 85 | 3546655 | ||
| BI823604.1 | - | 2 | 127 | |||
| D83200.1 | - | 1 | 33 | |||
| DA691216.1 | - | 2 | 133 | |||

URM1
Genomic Map