siRNA Details
| Gene Symbol: LBH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030915.3 | NP_112177.2 | 3 | 595 | ||
| XM_017005059.1 | XP_016860548.1 | 3 | 753 | |||
| GenBank mRNA | AK027232.1 | - | 3 | 383 | ||
| AK092206.1 | - | 3 | 1339 | |||
| AK223541.1 | - | 3 | 531 | |||
| AL136703.1 | - | 3 | 599 | |||
| BC109376.1 | - | 3 | 599 | 6495944 | ||
| BC126434.1 | - | 3 | 567 | 8992150 | ||
| BC130415.1 | - | 3 | 567 | 40146446 | ||
| EF025586.1 | - | 3 | 572 | |||

LBH
Genomic Map