siRNA Details
| Gene Symbol: LBH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030915.3 | NP_112177.2 | 3 | 1383 | ||
| XM_017005059.1 | XP_016860548.1 | 3 | 1541 | |||
| GenBank mRNA | AF110224.1 | - | 1 | 152 | ||
| AK023425.1 | - | 1 | 450 | |||
| AK027232.1 | - | 3 | 1175 | |||
| AK223541.1 | - | 3 | 1319 | |||
| AL136703.1 | - | 3 | 1388 | |||
| BC109376.1 | - | 3 | 1387 | 6495944 | ||
| BC126434.1 | - | 3 | 1356 | 8992150 | ||
| BC130415.1 | - | 3 | 1355 | 40146446 | ||
| EF025586.1 | - | 3 | 1361 | |||

LBH
Genomic Map