siRNA Details
| Gene Symbol: ACSBG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289177.1 | NP_001276106.1 | 3 | 568 | ||
| NM_001289178.1 | NP_001276107.1 | 3 | 514 | |||
| NM_001289179.1 | NP_001276108.1 | 3 | 559 | |||
| NM_001289180.1 | NP_001276109.1 | 4 | 399 | |||
| NM_001321384.1 | NP_001308313.1 | 3 | 549 | |||
| NM_030924.4 | NP_112186.3 | 3 | 559 | |||
| XM_005259653.3 | XP_005259710.1 | 3 | 541 | |||
| XM_017027332.1 | XP_016882821.1 | 3 | 386 | |||
| XM_017027333.1 | XP_016882822.1 | 3 | 541 | |||
| XM_017027334.1 | XP_016882823.1 | 3 | 541 | |||
| GenBank mRNA | AJ577571.1 | - | 3 | 462 | ||
| AK093433.1 | - | 3 | 527 | |||
| AK225862.1 | - | 2 | 267 | |||
| AK302604.1 | - | 3 | 516 | |||
| AL136854.1 | - | 3 | 300 | |||
| AY009107.1 | - | 4 | 392 | |||
| AY358766.1 | - | 3 | 559 | |||
| BC022027.1 | - | 3 | 490 | 4825681 | ||
| BC090046.1 | - | 3 | 475 | 30388900 | ||
| BI561348.1 | - | 3 | 520 | |||
| DC399703.1 | - | 4 | 399 | |||

ACSBG2
Genomic Map