siRNA Details
| Gene Symbol: ITM2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012514.2 | NP_001012532.1 | 2 | 354 | ||
| NM_001012516.2 | NP_001012534.1 | 3 | 440 | |||
| NM_001287240.1 | NP_001274169.1 | 3 | 431 | |||
| NM_001287241.1 | NP_001274170.1 | 4 | 627 | |||
| NM_030926.5 | NP_112188.1 | 3 | 495 | |||
| GenBank mRNA | AB003629.1 | - | 3 | 364 | ||
| AF271781.1 | - | 3 | 480 | |||
| AF272043.1 | - | 3 | 477 | |||
| AK056321.1 | - | 2 | 301 | |||
| AK075361.1 | - | 3 | 460 | |||
| AK090975.1 | - | 3 | 406 | |||
| AK222697.1 | - | 3 | 442 | |||
| AK295503.1 | - | 3 | 408 | |||
| AK307702.1 | - | 2 | 263 | |||
| AL136603.1 | - | 3 | 436 | |||
| AL713651.1 | - | 3 | 419 | |||
| AY049777.1 | - | 3 | 381 | |||
| BC002424.1 | - | 3 | 422 | 3162430 | ||
| BC025742.1 | - | 3 | 410 | 5224043 | ||
| BC050668.1 | - | 2 | 289 | 5759920 | ||
| BC098563.1 | - | 3 | 408 | 30558635 | ||
| BM842614.1 | - | 3 | 484 | |||
| BX444331.2 | - | 3 | 430 | |||
| CR457208.1 | - | 3 | 364 | |||
| DA138409.1 | - | 4 | 464 | |||
| DA182471.1 | - | 3 | 497 | |||

ITM2C
Genomic Map