siRNA Details
| Gene Symbol: UNC93B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030930.3 | NP_112192.2 | 4 | 490 | ||
| XM_011545290.1 | XP_011543592.1 | 2 | 100 | |||
| XM_011545291.2 | XP_011543593.1 | 3 | 312 | |||
| GenBank mRNA | AJ271326.1 | - | 4 | 449 | ||
| AJ422142.1 | - | 4 | 458 | |||
| AY007125.1 | - | 3 | 277 | |||
| BC025669.1 | - | 4 | 436 | |||
| BC033623.1 | - | 4 | 488 | |||
| BC092472.1 | - | 4 | 458 | 30348121 | ||
| BC101568.1 | - | 4 | 434 | 8069074 | ||
| BC105104.1 | - | 4 | 434 | 8144107 | ||

UNC93B1
Genomic Map