siRNA Details
| Gene Symbol: TSSK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052841.3 | NP_443073.1 | 1 | 504 | ||
| GenBank mRNA | AF296450.1 | - | 1 | 166 | ||
| AY048701.1 | - | 1 | 158 | |||
| BC035354.1 | - | 1 | 568 | 5171736 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_080442, AF298901, AK005983, AK019840, BC048470, and others.

TSSK3
Genomic Map