siRNA Details
| Gene Symbol: CYB561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017916.1 | NP_001017916.1 | 4 | 643 | ||
| NM_001017917.1 | NP_001017917.1 | 4 | 476 | |||
| NM_001915.3 | NP_001906.3 | 4 | 473 | |||
| XM_005257091.1 | XP_005257148.1 | 4 | 418 | |||
| XM_017024260.1 | XP_016879749.1 | 4 | 515 | |||
| XM_017024261.1 | XP_016879750.1 | 3 | 356 | |||
| XM_017024262.1 | XP_016879751.1 | 4 | 708 | |||
| GenBank mRNA | AK095244.1 | - | 4 | 690 | ||
| AK294403.1 | - | 4 | 410 | |||
| AK294630.1 | - | 4 | 627 | |||
| AK301541.1 | - | 4 | 403 | |||
| AK303177.1 | - | 4 | 342 | |||
| AK316606.1 | - | 4 | 418 | |||
| BC000021.1 | - | 4 | 504 | 3509626 | ||
| BC002976.1 | - | 4 | 430 | 3535771 | ||
| BC091485.1 | - | 4 | 444 | 6188259 | ||
| BF206825.1 | - | 478 | ||||
| BM552101.1 | - | 4 | 473 | |||
| BT007096.1 | - | 3 | 343 | |||

CYB561
Genomic Map