siRNA Details
| Gene Symbol: SLIRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267864.1 | NP_001254793.1 | 4 | 329 | ||
| NM_031210.5 | NP_112487.1 | 3, 4 | 313 | |||
| NR_052025.1 | 4, 5 | 424 | ||||
| GenBank mRNA | AF253980.1 | - | 3, 4 | 284 | ||
| AF271779.1 | - | 3, 4 | 308 | |||
| AJ272055.1 | - | 3, 4 | 272 | |||
| AY860853.1 | - | 3, 4 | 272 | |||
| BC017895.1 | - | 3, 4 | 281 | 4250073 | ||
| BM685909.1 | - | 4, 5 | 389 | |||
| BP289922.1 | - | 4 | 301 | |||
| BQ083727.1 | - | 4 | 320 | |||
| BU944910.1 | - | 3, 4 | 313 | |||
| BU960115.1 | - | 3, 4 | 291 | |||
| EL951262.1 | - | 3, 4 | 273 | |||

SLIRP
Genomic Map