siRNA Details
| Gene Symbol: ZNF93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031218.3 | NP_112495.2 | 4 | 1410 | ||
| GenBank mRNA | AK022550.1 | - | 1 | 317 | ||
| AK295921.1 | - | 1 | 742 | |||
| BC039001.1 | - | 1 | 386 | |||
| BC136718.1 | - | 4 | 1262 | 9020708 | ||
| BC136719.1 | - | 4 | 1262 | 9020709 | ||

ZNF93
Genomic Map