siRNA Details
| Gene Symbol: PITPNM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165966.1 | NP_001159438.1 | 4 | 377 | ||
| NM_031220.3 | NP_112497.2 | 5 | 485 | |||
| XM_011524015.2 | XP_011522317.1 | 5 | 425 | |||
| XM_011524016.2 | XP_011522318.1 | 5 | 426 | |||
| XM_011524017.2 | XP_011522319.1 | 5 | 427 | |||
| GenBank mRNA | AF334586.1 | - | 5 | 482 | ||
| AL389994.1 | - | 4 | 208 | |||
| BC128583.1 | - | 5 | 422 | 40128716 | ||
| BC128584.1 | - | 5 | 422 | 40128724 | ||

PITPNM3
Genomic Map