siRNA Details
| Gene Symbol: CYC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001916.4 | NP_001907.2 | 6 | 852 | ||
| XM_017013102.1 | XP_016868591.1 | 5 | 1385 | |||
| GenBank mRNA | AK026633.1 | - | 6 | 810 | ||
| AK225796.1 | - | 5 | 1453 | |||
| BC001006.1 | - | 6 | 798 | 2821836 | ||
| BC015616.2 | - | 6 | 792 | 4840922 | ||
| BC020566.1 | - | 6 | 791 | 4157475 | ||
| BC024650.1 | - | 6 | 759 | |||
| BT019798.1 | - | 6 | 787 | |||
| CR541674.1 | - | 6 | 787 | |||
| M16597.1 | - | 5 | 493 | |||
| X06994.1 | - | 6 | 795 | |||

CYC1
Genomic Map