siRNA Details
| Gene Symbol: CYLC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001340.4 | NP_001331.1 | 4, 5 | 410 | ||
| GenBank mRNA | AK313349.1 | - | 4, 5 | 397 | ||
| BC058038.1 | - | 4, 5 | 414 | |||
| BC114547.1 | - | 4, 5 | 343 | 40005379 | ||
| Z46788.1 | - | 4, 5 | 343 | |||

CYLC2
Genomic Map