siRNA Details
| Gene Symbol: RTP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031440.1 | NP_113628.1 | 2 | 330 | ||
| GenBank mRNA | AJ312776.1 | - | 2 | 330 | ||
| AY562237.1 | - | 2 | 262 | |||
| BC131756.1 | - | 2 | 291 | 40119315 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RTP3| Gene Symbol: RTP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031440.1 | NP_113628.1 | 2 | 330 | ||
| GenBank mRNA | AJ312776.1 | - | 2 | 330 | ||
| AY562237.1 | - | 2 | 262 | |||
| BC131756.1 | - | 2 | 291 | 40119315 | ||