siRNA Details
| Gene Symbol: CCM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029835.2 | NP_001025006.1 | 2 | 321 | ||
| NM_001167935.1 | NP_001161407.1 | 2 | 333 | |||
| NM_031443.3 | NP_113631.1 | 2 | 333 | |||
| NR_030770.1 | 2 | 269 | ||||
| XM_006715786.3 | XP_006715849.1 | 2 | 929 | |||
| XM_011515561.2 | XP_011513863.1 | 2 | 283 | |||
| XM_011515562.1 | XP_011513864.1 | 2 | 314 | |||
| XM_017012671.1 | XP_016868160.1 | 2 | 283 | |||
| GenBank mRNA | AF370392.1 | - | 2 | 333 | ||
| AK025807.1 | - | 1 | 48 | |||
| AK098005.1 | - | 2 | 929 | |||
| AK298671.1 | - | 2 | 283 | |||
| AK303534.1 | - | 2 | 285 | |||
| BC004903.2 | - | 2 | 268 | 3501896 | ||
| BC008859.2 | - | 2 | 268 | 4124185 | ||
| BC016832.2 | - | 2 | 262 | 2924210 | ||
| BC063663.1 | - | 2 | 268 | |||
| DA420491.1 | - | 2 | 257 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_146014, AK206057, BC029157, AY442689, AK155145.

CCM2
Genomic Map