siRNA Details
| Gene Symbol: ZMIZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300959.1 | NP_001287888.1 | 8 | 1113 | ||
| NM_031449.3 | NP_113637.3 | 9 | 1282 | |||
| NM_174929.2 | NP_777589.2 | 7 | 1143 | |||
| XM_005249866.2 | XP_005249923.1 | 9 | 1306 | |||
| XM_005249867.4 | XP_005249924.1 | 9 | 1308 | |||
| XM_005249868.4 | XP_005249925.1 | 9 | 2326 | |||
| XM_005249869.2 | XP_005249926.1 | 9 | 1306 | |||
| XM_005249870.2 | XP_005249927.1 | 8 | 1228 | |||
| XM_005249871.2 | XP_005249928.1 | 9 | 1210 | |||
| XM_005249872.2 | XP_005249929.1 | 8 | 1228 | |||
| XM_005249873.2 | XP_005249930.1 | 9 | 1210 | |||
| XM_011515566.1 | XP_011513868.1 | 3 | 446 | |||
| XM_017012674.1 | XP_016868163.1 | 9 | 1387 | |||
| GenBank mRNA | AB067473.1 | - | 3 | 366 | ||
| AF357908.1 | - | 8 | 1159 | |||
| AK090415.1 | - | 9 | 1282 | |||
| AK090841.1 | - | 1 | 562 | |||
| AL831933.1 | - | 1 | 779 | |||
| AY426594.1 | - | 7 | 1105 | |||
| BC016968.1 | - | 7 | 1126 | |||
| BC021924.1 | - | 3 | 214 | |||
| BC110435.1 | - | 8 | 1098 | 5518312 | ||

ZMIZ2
Genomic Map