siRNA Details
| Gene Symbol: FAM103A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031452.3 | NP_113640.1 | 4 | 455 | ||
| GenBank mRNA | AK027095.1 | - | 4 | 416 | ||
| AL049237.1 | - | 1 | 20 | |||
| BC003627.2 | - | 1 | 415 | 2989772 | ||
| BC027181.1 | - | 1 | 407 | 3924570 | ||
| BC112329.1 | - | 1 | 424 | 5578103 | ||

FAM103A1
Genomic Map