siRNA Details
| Gene Symbol: PARP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146102.1 | NP_001139574.1 | 3 | 249 | ||
| NM_031458.2 | NP_113646.2 | 3 | 431 | |||
| XM_005247820.2 | XP_005247877.1 | 3 | 355 | |||
| XM_011513218.1 | XP_011511520.1 | 3 | 221 | |||
| XM_017007304.1 | XP_016862793.1 | 3 | 239 | |||
| GenBank mRNA | AF307338.1 | - | 3 | 332 | ||
| AK313494.1 | - | 3 | 218 | |||

PARP9
Genomic Map