siRNA Details
| Gene Symbol: CYP3A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000765.4 | NP_000756.3 | 11 | 1218 | ||
| GenBank mRNA | BC067436.1 | - | 11 | 1196 | 7002135 | |
| BX647143.1 | - | 8 | 1450 | |||
| D00408.1 | - | 11 | 1114 | |||
| Gene Symbol: CYP3A7-CYP3A51P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256497.2 | NP_001243426.2 | 11 | 1218 | ||
| GenBank mRNA | AF315325.1 | - | 11 | 1111 | ||

CYP3A7
Genomic Map