siRNA Details
Gene Symbol: CYP3A7-CYP3A51P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256497.2 | NP_001243426.2 | 7 | 768 | ||
GenBank mRNA | AF315325.1 | - | 7 | 661 |
Gene Symbol: CYP3A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000765.4 | NP_000756.3 | 7 | 768 | ||
GenBank mRNA | BC067436.1 | - | 7 | 746 | 7002135 | |
BX647143.1 | - | 4 | 1000 | |||
D00408.1 | - | 7 | 664 |
