siRNA Details
| Gene Symbol: FAM57B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318504.1 | NP_001305433.1 | 2, 3 | 102 | ||
| NM_031478.5 | NP_113666.2 | 2, 3 | 540 | |||
| XM_005255613.3 | XP_005255670.1 | 6, 7 | 748 | |||
| XM_005255614.3 | XP_005255671.1 | 5, 6 | 688 | |||
| XM_005255615.2 | XP_005255672.1 | 5, 6 | 885 | |||
| XM_017023749.1 | XP_016879238.1 | 3, 4 | 732 | |||
| XM_017023750.1 | XP_016879239.1 | 6, 7 | 945 | |||
| XM_017023751.1 | XP_016879240.1 | 7, 8 | 841 | |||
| XM_017023752.1 | XP_016879241.1 | 6, 7 | 744 | |||
| XM_017023753.1 | XP_016879242.1 | 4, 5 | 824 | |||
| XM_017023754.1 | XP_016879243.1 | 3, 4 | 246 | |||
| GenBank mRNA | AB593089.1 | - | 2, 3 | 480 | ||
| AB593090.1 | - | 2, 3 | 478 | |||
| AF370365.1 | - | 2, 3 | 492 | |||
| AL136777.1 | - | 2, 3 | 102 | |||
| AL535138.3 | - | 2, 3 | 548 | |||
| BC007892.2 | - | 2, 3 | 258 | 4126084 | ||

FAM57B
Genomic Map