siRNA Details
| Gene Symbol: INHBE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031479.3 | NP_113667.1 | 2 | 1227 | ||
| GenBank mRNA | AF412024.1 | - | 2 | 1211 | ||
| AK075285.1 | - | 2 | 1222 | |||
| AK308647.1 | - | 2 | 922 | |||
| BC005161.2 | - | 2 | 1219 | 3529092 | ||

INHBE
Genomic Map