siRNA Details
| Gene Symbol: LONP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300948.1 | NP_001287877.1 | 2 | 663 | ||
| NM_031490.3 | NP_113678.2 | 2 | 663 | |||
| XM_017023755.1 | XP_016879244.1 | 2 | 530 | |||
| XM_017023756.1 | XP_016879245.1 | 2 | 528 | |||
| GenBank mRNA | AJ548761.1 | - | 2 | 530 | ||
| AK027544.1 | - | 2 | 494 | |||
| AK074775.1 | - | 2 | 528 | |||
| AK127125.1 | - | 3 | 426 | |||
| AK294779.1 | - | 2 | 475 | |||
| AL834201.1 | - | 1 | 30 | |||
| BC093910.1 | - | 2 | 497 | 7939755 | ||
| BC093912.1 | - | 2 | 497 | 7939757 | ||
| BC143246.1 | - | 2 | 497 | 9051747 | ||

LONP2
Genomic Map