siRNA Details
| Gene Symbol: HMCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031935.2 | NP_114141.2 | 3 | 663 | ||
| XM_011510038.2 | XP_011508340.1 | 3 | 680 | |||
| XM_011510041.2 | XP_011508343.1 | 3 | 688 | |||
| GenBank mRNA | AF156100.1 | - | 3 | 663 | ||

HMCN1
Genomic Map