siRNA Details
| Gene Symbol: HMCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031935.2 | NP_114141.2 | 12 | 2167 | ||
| XM_011510038.2 | XP_011508340.1 | 12 | 2184 | |||
| XM_011510041.2 | XP_011508343.1 | 12 | 2192 | |||
| XM_017002437.1 | XP_016857926.1 | 1 | 110 | |||
| GenBank mRNA | AF156100.1 | - | 12 | 2167 | ||
| AK056336.1 | - | 2 | 182 | |||

HMCN1
Genomic Map