siRNA Details
| Gene Symbol: GPR61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031936.4 | NP_114142.3 | 3 | 2538 | ||
| XM_017002438.1 | XP_016857927.1 | 2 | 5542 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR61| Gene Symbol: GPR61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031936.4 | NP_114142.3 | 3 | 2538 | ||
| XM_017002438.1 | XP_016857927.1 | 2 | 5542 | |||
Set Membership: |
|