siRNA Details
| Gene Symbol: TBC1D10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204240.1 | NP_001191169.1 | 9 | 1287 | ||
| NM_031937.2 | NP_114143.1 | 9 | 1266 | |||
| GenBank mRNA | AB449894.1 | - | 9 | 1181 | ||
| AF331038.1 | - | 9 | 1205 | |||
| AK074656.1 | - | 9 | 1232 | |||
| AK093231.1 | - | 6 | 2401 | |||
| AK127939.1 | - | 9 | 1222 | |||
| AK131086.1 | - | 5 | 4831 | |||
| AK309455.1 | - | 9 | 1032 | |||
| BC022201.1 | - | 9 | 1215 | |||
| BC136815.1 | - | 9 | 1222 | 9020806 | ||
| BC142940.1 | - | 9 | 1238 | 8860306 | ||
| BC150187.1 | - | 9 | 1186 | 40148357 | ||
| BC150214.1 | - | 9 | 1191 | 40149007 | ||
| BC171775.1 | - | 9 | 1222 | 9054429 | ||
| CT841514.1 | - | 9 | 1187 | |||

TBC1D10A
Genomic Map