siRNA Details
| Gene Symbol: KRTAP3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031958.1 | NP_114164.1 | 1 | 559 | ||
| GenBank mRNA | AJ406931.1 | - | 1 | 559 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP3-1| Gene Symbol: KRTAP3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031958.1 | NP_114164.1 | 1 | 559 | ||
| GenBank mRNA | AJ406931.1 | - | 1 | 559 | ||