siRNA Details
| Gene Symbol: STK40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282546.1 | NP_001269475.1 | 5 | 616 | ||
| NM_001282547.1 | NP_001269476.1 | 5 | 601 | |||
| NM_032017.2 | NP_114406.1 | 6 | 838 | |||
| GenBank mRNA | AK024504.1 | - | 4 | 400 | ||
| AK075029.1 | - | 2 | 117 | |||
| AK075048.1 | - | 5 | 598 | |||
| AL834137.1 | - | 6 | 795 | |||
| AY198395.1 | - | 5 | 571 | |||
| BC007835.2 | - | 5 | 569 | 4109652 | ||
| BC008344.1 | - | 5 | 577 | 3530483 | ||

STK40
Genomic Map