siRNA Details
| Gene Symbol: HDAC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159286.1 | NP_001152758.1 | 16 | 1934 | ||
| NM_032019.5 | NP_114408.3 | 17 | 1994 | |||
| GenBank mRNA | AF393962.1 | - | 17 | 1621 | ||
| AF407272.1 | - | 17 | 1621 | |||
| AF407273.1 | - | 16 | 1561 | |||
| AF426160.1 | - | 17 | 1974 | |||
| AY450395.1 | - | 14 | 1386 | |||
| BC032049.1 | - | 17 | 1807 | |||
| BC094734.1 | - | 15 | 1662 | |||
| BC125082.1 | - | 11 | 1143 | |||
| BC125083.1 | - | 17 | 1668 | 40117772 | ||
| CR456465.1 | - | 17 | 1677 | |||

HDAC10
Genomic Map