siRNA Details
| Gene Symbol: TMEM133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032021.2 | NP_114410.1 | 1 | 431 | ||
| GenBank mRNA | AF247167.1 | - | 1 | 431 | ||
| AK314174.1 | - | 1 | 522 | |||
| BC093843.1 | - | 1 | 277 | 7939688 | ||
| BC112050.1 | - | 1 | 277 | 8327518 | ||
| Gene Symbol: ARHGAP42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011542615.2 | XP_011540917.1 | 24 | 8825 | ||
| XM_011542616.2 | XP_011540918.1 | 24 | 6594 | |||
| XM_011542617.2 | XP_011540919.1 | 25 | 6480 | |||
| XM_017017237.1 | XP_016872726.1 | 24 | 6783 | |||
| XM_017017238.1 | XP_016872727.1 | 25 | 8966 | |||

TMEM133
Genomic Map