siRNA Details
| Gene Symbol: SPNS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142448.1 | NP_001135920.1 | 8, 9 | 1105 | ||
| NM_001142450.1 | NP_001135922.1 | 8, 9 | 1255 | |||
| NM_032038.2 | NP_114427.1 | 7, 8 | 1342 | |||
| XM_017023757.1 | XP_016879246.1 | 8, 9 | 1128 | |||
| XM_017023758.1 | XP_016879247.1 | 8, 9 | 1124 | |||
| XM_017023759.1 | XP_016879248.1 | 8, 9 | 1108 | |||
| GenBank mRNA | AF212371.1 | - | 7, 8 | 1289 | ||
| AF370423.1 | - | 8, 9 | 1237 | |||
| BC006156.1 | - | 7, 8 | 953 | |||
| BC008325.1 | - | 7, 8 | 1167 | 3501826 | ||
| BC038961.1 | - | 8, 9 | 1119 | 6053095 | ||
| BC065235.1 | - | 8, 9 | 1087 | 6161778 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_023712, AK199707, BC085491, AK195969, AK166931, and others.

SPNS1
Genomic Map