siRNA Details
| Gene Symbol: CCDC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032040.4 | NP_114429.2 | 1 | 2275 | ||
| GenBank mRNA | AK297668.1 | - | 2 | 1722 | ||
| AL136609.1 | - | 1 | 2201 | |||
| BC025243.1 | - | 1 | 1646 | 4553965 | ||
| JN703457.1 | - | 1 | 2197 | |||
| KT894208.1 | - | 1 | 1431 | |||

CCDC8
Genomic Map