siRNA Details
| Gene Symbol: TMPRSS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077263.2 | NP_001070731.1 | 11 | 1607 | ||
| NM_001206789.1 | NP_001193718.1 | 10 | 1502 | |||
| NM_001244995.1 | NP_001231924.1 | 11 | 1607 | |||
| GenBank mRNA | AB048796.1 | - | 11 | 1533 | ||
| AB048797.1 | - | 10 | 1458 | |||
| AK300283.1 | - | 11 | 1519 | |||
| AY190317.1 | - | 11 | 1534 | |||
| BC018715.1 | - | 12 | 1342 | |||
| BC114928.1 | - | 11 | 1526 | 40028749 | ||

TMPRSS13
Genomic Map