siRNA Details
| Gene Symbol: KATNAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014380.2 | NP_001014402.1 | 7 | 951 | ||
| NM_032116.4 | NP_115492.1 | 7 | 993 | |||
| XM_005266574.2 | XP_005266631.1 | 7 | 1135 | |||
| XM_017020791.1 | XP_016876280.1 | 6 | 837 | |||
| XM_017020792.1 | XP_016876281.1 | 6 | 858 | |||
| XM_017020793.1 | XP_016876282.1 | 4 | 1167 | |||
| GenBank mRNA | AK291439.1 | - | 7 | 926 | ||
| AK315724.1 | - | 7 | 930 | |||
| BC000612.2 | - | 7 | 953 | 3347282 | ||

KATNAL1
Genomic Map