siRNA Details
| Gene Symbol: MND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032117.3 | NP_115493.1 | 6 | 456 | ||
| NR_045605.1 | 8 | 617 | ||||
| XM_005263275.2 | XP_005263332.1 | 6 | 415 | |||
| XM_017008688.1 | XP_016864177.1 | 6 | 813 | |||
| GenBank mRNA | AK312172.1 | - | 6 | 415 | ||
| AY028916.1 | - | 6 | 395 | |||
| BC032142.2 | - | 6 | 410 | 5095005 | ||
| BF975093.1 | - | 8 | 585 | |||
| BG758047.1 | - | 8 | 586 | |||
| BU533354.1 | - | 3 | 112 | |||

MND1
Genomic Map