siRNA Details
Gene Symbol: TMEM222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032125.2 | NP_115501.2 | 5 | 513 | ||
NR_037576.1 | 6 | 666 | ||||
NR_037577.1 | 5 | 513 | ||||
NR_037580.1 | 6 | 666 | ||||
GenBank mRNA | AF251699.1 | - | 5 | 409 | ||
AK095889.1 | - | 2 | 645 | |||
AK123218.1 | - | 2 | 1761 | |||
AK222642.1 | - | 5 | 392 | |||
AK303821.1 | - | 5 | 467 | |||
AK307077.1 | - | 6 | 504 | |||
AK313110.1 | - | 5 | 495 | |||
AL136683.1 | - | 5 | 427 | |||
BC011579.1 | - | 5 | 410 | |||
BC090039.1 | - | 6 | 558 | 6181849 | ||
BP326836.1 | - | 5 | 498 | |||
BQ927627.1 | - | 6 | 471 | |||
BX394857.2 | - | 5 | 373 | |||
DC424669.1 | - | 5 | 515 |
