siRNA Details
| Gene Symbol: TMEM222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032125.2 | NP_115501.2 | 6 | 1106 | ||
| NR_037576.1 | 7 | 1259 | ||||
| NR_037577.1 | 7 | 1194 | ||||
| NR_037580.1 | 8 | 1347 | ||||
| GenBank mRNA | AK095889.1 | - | 2 | 2335 | ||
| AK123218.1 | - | 3 | 2354 | |||
| AK222642.1 | - | 6 | 985 | |||
| AK303821.1 | - | 6 | 1060 | |||
| AL136683.1 | - | 6 | 1020 | |||
| BC011579.1 | - | 6 | 1003 | |||
| BC090039.1 | - | 7 | 1151 | 6181849 | ||

TMEM222
Genomic Map