siRNA Details
| Gene Symbol: SLC10A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029998.5 | NP_001025169.1 | 2 | 422 | ||
| NM_001300842.2 | NP_001287771.1 | 2 | 422 | |||
| NM_001317816.1 | NP_001304745.1 | 2 | 422 | |||
| NM_001317817.1 | NP_001304746.1 | 2 | 422 | |||
| NM_001317818.1 | NP_001304747.1 | 2 | 422 | |||
| NM_032128.4 | NP_115504.1 | 2 | 422 | |||
| NR_133924.1 | 2 | 422 | ||||
| XM_011532311.2 | XP_011530613.1 | 2 | 422 | |||
| XM_011532313.2 | XP_011530615.1 | 1 | 68 | |||
| XM_017008689.1 | XP_016864178.1 | 2 | 422 | |||
| XM_017008690.1 | XP_016864179.1 | 2 | 422 | |||
| XM_017008691.1 | XP_016864180.1 | 2 | 422 | |||
| XM_017008692.1 | XP_016864181.1 | 2 | 802 | |||
| GenBank mRNA | AK291046.1 | - | 2 | 290 | ||
| AL136728.1 | - | 2 | 350 | |||
| AY346324.1 | - | 2 | 385 | |||
| BC023288.1 | - | 2 | 373 | |||
| BC063471.1 | - | 2 | 335 | |||
| BC120970.1 | - | 2 | 281 | |||
| BC120971.1 | - | 2 | 281 | |||
| BC127626.1 | - | 2 | 281 | |||
| BC127627.1 | - | 2 | 281 | |||
| BC148252.1 | - | 2 | 281 | |||
| BC150308.1 | - | 2 | 391 | 8860437 | ||
| CR933647.1 | - | 2 | 416 | |||
| CR936751.1 | - | 2 | 279 | |||
| DA590625.1 | - | 2 | 423 | |||
| DB205503.1 | - | 2 | 422 | |||
| DQ122860.1 | - | 2 | 168 | |||
| DQ122861.1 | - | 2 | 168 | |||
| DQ871036.1 | - | 2 | 168 | |||

SLC10A7
Genomic Map