siRNA Details
| Gene Symbol: PLEKHN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160184.1 | NP_001153656.1 | 15 | 2165 | ||
| NM_032129.2 | NP_115505.2 | 16 | 2270 | |||
| GenBank mRNA | AL136730.1 | - | 16 | 2270 | ||
| BC101386.2 | - | 15 | 2160 | 40026400 | ||
| BC101387.2 | - | 15 | 2421 | 40026404 | ||

PLEKHN1
Genomic Map