siRNA Details
| Gene Symbol: C3orf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184957.1 | NP_001171886.1 | 12 | 1740 | ||
| NM_001184958.1 | NP_001171887.1 | 12 | 1845 | |||
| NM_032137.4 | NP_115513.4 | 12 | 2242 | |||
| XM_006713341.2 | XP_006713404.1 | 12 | 2242 | |||
| XM_006713342.2 | XP_006713405.1 | 12 | 2095 | |||
| XM_006713343.2 | XP_006713406.1 | 12 | 2242 | |||
| XM_011534153.2 | XP_011532455.1 | 12 | 3872 | |||
| XM_011534154.2 | XP_011532456.1 | 12 | 3307 | |||
| XM_011534157.2 | XP_011532459.1 | 8 | 1019 | |||
| XM_011534158.2 | XP_011532460.1 | 11 | 2118 | |||
| GenBank mRNA | AK131481.1 | - | 13 | 2448 | ||
| AL136781.1 | - | 12 | 1660 | |||
| AL834386.1 | - | 12 | 2194 | |||
| BC030599.2 | - | 12 | 1803 | 4831210 | ||
| BC038406.1 | - | 12 | 2162 | 5167835 | ||

C3orf20
Genomic Map