siRNA Details
| Gene Symbol: C3orf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL834386.1 | - | 8 | 1672 | ||
| BC030599.2 | - | 8 | 1281 | 4831210 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C3orf20| Gene Symbol: C3orf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL834386.1 | - | 8 | 1672 | ||
| BC030599.2 | - | 8 | 1281 | 4831210 | ||