siRNA Details
| Gene Symbol: FBXO30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032145.4 | NP_115521.3 | 2 | 804 | ||
| XM_005267159.3 | XP_005267216.1 | 2 | 643 | |||
| XM_011536177.2 | XP_011534479.1 | 2 | 5663 | |||
| XM_017011353.1 | XP_016866842.1 | 2 | 643 | |||
| GenBank mRNA | BC024326.1 | - | 2 | 772 | 3915249 | |

FBXO30
Genomic Map