siRNA Details
| Gene Symbol: CYP3A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000777.4 | NP_000768.1 | 8, 9 | 936 | ||
| NM_001291829.1 | NP_001278758.1 | 9, 10 | 1436 | |||
| NM_001291830.1 | NP_001278759.1 | 9, 10 | 1196 | |||
| XM_006715859.2 | XP_006715922.1 | 8, 9 | 903 | |||
| XM_011515843.1 | XP_011514145.1 | 9, 10 | 1053 | |||
| XM_017011781.1 | XP_016867270.1 | 5, 6 | 578 | |||
| XM_017011782.1 | XP_016867271.1 | 3, 4 | 375 | |||
| GenBank mRNA | AK223008.1 | - | 8, 9 | 906 | ||
| AK296205.1 | - | 9, 10 | 1163 | |||
| AK313813.1 | - | 8, 9 | 903 | |||
| BC033862.1 | - | 8, 9 | 895 | 5188554 | ||
| J04813.1 | - | 8, 9 | 893 | |||
| L26985.1 | - | 10, 11 | 1126 | |||
| X90579.1 | - | 9, 10 | 1025 | |||

CYP3A5
Genomic Map