siRNA Details
| Gene Symbol: LRRC8C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032270.4 | NP_115646.2 | 3 | 1456 | ||
| XM_006710960.3 | XP_006711023.1 | 3 | 3098 | |||
| XM_011542282.2 | XP_011540584.1 | 3 | 1443 | |||
| XM_011542283.2 | XP_011540585.1 | 4 | 3010 | |||
| XM_017002503.1 | XP_016857992.1 | 3 | 1303 | |||
| XM_017002504.1 | XP_016857993.1 | 4 | 1212 | |||
| GenBank mRNA | AB081509.1 | - | 3 | 1337 | ||
| AK127881.1 | - | 3 | 1393 | |||
| AL136919.1 | - | 1 | 956 | |||
| BC113973.1 | - | 3 | 1263 | 40007964 | ||

LRRC8C
Genomic Map